Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012055.3 | NP_036185.1 | 5 - 6 | 876 | 67 | |
GenBank mRNA | AK076207.1 | - | 5 - 6 | 894 | 67 | |
AK160459.1 | - | 5 - 6 | 891 | 67 | ||
AK167524.1 | - | 5 - 6 | 856 | 67 | ||
BC005552.1 | - | 5 - 6 | 874 | 3585775 | 67 | |
BC050123.1 | - | 3204 | 67 | |||
U38940.1 | - | 5 - 6 | 877 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |