Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23542444 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23542440 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23542439 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23542442 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23542441 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2079483 |
AY319256.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2079483 |
NM_015739.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2079483 |
AF547996.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2051787 |
NM_015739.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2051787 |
AF547996.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
656657 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
463479 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
475148 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
81541 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
945867 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
581166 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
70236 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
134252 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
805053 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
591511 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
54416 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
782140 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
602041 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454989 |
AK016491.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454989 |
AK045911.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454989 |
AK144175.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454989 |
AK171846.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454989 |
BC019531.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454989 |
BC085088.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454989 |
NM_028889.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
153245 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
100581 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P105252 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P133792 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2079483 |
AY319256.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2079483 |
NM_015739.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2079483 |
AF547996.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2051787 |
NM_015739.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2051787 |
AF547996.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P459811 |
AY319256.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P459811 |
NM_015739.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P459811 |
AF547996.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542438 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542435 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542469 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542461 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542474 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542431 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542473 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542463 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542428 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542451 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542465 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542482 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542466 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542425 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542468 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542470 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542487 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542485 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542486 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542454 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542458 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542442 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542477 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542481 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542489 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542479 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542439 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542467 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542453 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542462 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542433 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542478 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542484 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542456 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542426 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542476 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P23542490 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05507236 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05507208 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P133792 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05507200 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P105252 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P459811 |
AY319256.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P459811 |
NM_015739.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P459811 |
AF547996.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P123870 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P110768 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P105252 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P05507200 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P133792 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04516706 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04597189 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04652550 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04652551 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04652553 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23542442 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23542440 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23542439 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23542444 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P23542441 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542461 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542484 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542474 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542509 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542330 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542402 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542453 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542372 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542358 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542517 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542411 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542387 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542376 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542469 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542423 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542379 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542315 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542426 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542465 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542504 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542408 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542329 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542458 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542412 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542343 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542313 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542360 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542322 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542415 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542320 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542489 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542364 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542354 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542420 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542454 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542421 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542328 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542356 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542476 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542339 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542470 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542485 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542438 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542317 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542482 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542352 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542399 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542382 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542466 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542344 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542393 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542428 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542467 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542391 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542375 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542324 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542395 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542337 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542419 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542371 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542431 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542407 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542451 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542439 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542478 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542424 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542332 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542435 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542456 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542487 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542367 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542389 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542403 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542378 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542463 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542380 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542416 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542341 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542327 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542433 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542350 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542335 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542418 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542377 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P23542481 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04248947 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542421 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542435 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542405 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542438 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542324 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542428 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04533488 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04248944 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542356 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542439 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542315 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542481 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542330 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542458 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04246343 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542398 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542402 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542418 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542485 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542360 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542415 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542362 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04248951 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542454 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542317 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542470 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542478 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04102397 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542426 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04102381 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04533490 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542413 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04699812 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542399 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04699814 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542414 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542433 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542411 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542351 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04511086 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542486 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542461 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542403 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542354 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542463 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04856832 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542487 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542327 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542474 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542409 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04506503 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542328 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04663993 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542322 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542451 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04506496 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542407 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04246345 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542453 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04763266 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542476 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542320 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542352 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542436 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542467 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P23542456 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04699791 |
- |
- |