Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026791.2 | NP_081067.2 | 6 - 7 | 1045 | 63 | |
GenBank mRNA | AK003747.1 | - | 1 - 2 | 109 | 63 | |
AK082085.1 | - | 5 - 6 | 1534 | 63 | ||
AK156709.1 | - | 5 - 6 | 1195 | 63 | ||
BC043658.1 | - | 6 - 7 | 1006 | 63 | ||
BC053694.1 | - | 4 - 5 | 461 | 63 | ||
BC060082.1 | - | 3 - 4 | 276 | 63 | ||
BC144773.1 | - | 6 - 7 | 1411 | 63 | ||
BC145661.1 | - | 6 - 7 | 1411 | 63 | ||
CF197934.1 | - | 4 - 5 | 449 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |