Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027373.2 | NP_081649.1 | 5 - 6 | 769 | 58 | |
GenBank mRNA | AK011153.1 | - | 5 - 6 | 597 | 58 | |
AK080756.1 | - | 5 - 6 | 704 | 58 | ||
AK129483.1 | - | 5 - 6 | 696 | 58 | ||
AK147706.1 | - | 5 - 6 | 661 | 58 | ||
AK148000.1 | - | 5 - 6 | 624 | 58 | ||
AK161385.1 | - | 5 - 6 | 720 | 58 | ||
BC050814.1 | - | 5 - 6 | 769 | 6773872 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |