Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029278.2 | NP_083554.2 | 2 - 3 | 431 | 116 | |
GenBank mRNA | AK011648.1 | - | 2 - 3 | 407 | 116 | |
AK078605.1 | - | 2 - 3 | 417 | 116 | ||
AK079609.1 | - | 2 - 3 | 433 | 116 | ||
AK148605.1 | - | 2 - 3 | 390 | 116 | ||
AK161728.1 | - | 2 - 3 | 369 | 116 | ||
BC024998.1 | - | 2 - 3 | 352 | 5012070 | 116 | |
BC043043.1 | - | 2 - 3 | 374 | 5701430 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |