Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyw3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168358.1 | NP_001161830.1 | 2 - 3 | 327 | 94 | |
NM_172474.5 | NP_766062.1 | 2 - 3 | 327 | 94 | ||
GenBank mRNA | AK030344.1 | - | 2 - 3 | 325 | 94 | |
AK034820.1 | - | 2 - 3 | 312 | 94 | ||
BC086686.1 | - | 2 - 3 | 301 | 30680486 | 94 | |
BC087879.1 | - | 2 - 3 | 291 | 6533208 | 94 | |
BC141040.1 | - | 2 - 3 | 306 | 9055652 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |