Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136081.2 | NP_001129553.1 | 2 - 3 | 252 | 79 | |
NM_182990.4 | NP_892035.2 | 2 - 3 | 381 | 79 | ||
GenBank mRNA | AK146446.1 | - | 2 - 3 | 304 | 79 | |
AK146585.1 | - | 2 - 3 | 301 | 79 | ||
AK146607.1 | - | 2 - 3 | 284 | 79 | ||
AK146691.1 | - | 2 - 3 | 284 | 79 | ||
AK147005.1 | - | 2 - 3 | 314 | 79 | ||
AK151584.1 | - | 2 - 3 | 334 | 79 | ||
AK158552.1 | - | 2 - 3 | 252 | 79 | ||
BC004848.1 | - | 2 - 3 | 248 | 79 | ||
BC042502.1 | - | 2 - 3 | 229 | 5005209 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |