Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf2bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183029.2 | NP_898850.2 | 1 - 2 | 513 | 104 | |
GenBank mRNA | AK049196.1 | - | 1 - 2 | 210 | 104 | |
AK077404.1 | - | 1 - 2 | 192 | 104 | ||
AK159654.1 | - | 1 - 2 | 248 | 104 | ||
AY531659.1 | - | 1 - 2 | 177 | 104 | ||
BC023758.1 | - | 1 - 2 | 211 | 104 | ||
BC054552.1 | - | 1 - 2 | 233 | 104 | ||
CB290059.1 | - | 1 - 2 | 513 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |