Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spink8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183136.2 | NP_898959.2 | 2 - 3 | 254 | 128 | |
GenBank mRNA | AK021276.1 | - | 2 - 3 | 237 | 128 | |
BC048588.1 | - | 2 - 3 | 254 | 6707477 | 128 | |
CN843460.1 | - | 2 - 3 | 200 | 128 | ||
DQ437329.1 | - | 2 - 3 | 236 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |