Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183285.3 | NP_899108.3 | 2 - 3 | 491 | 87 | |
GenBank mRNA | AA537633.1 | - | 2 - 3 | 145 | 87 | |
AK009318.1 | - | 2 - 3 | 87 | |||
BC095988.1 | - | 2 - 3 | 450 | 5365458 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |