Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10607961 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10607959 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10607954 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10607957 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10607953 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10607955 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10607960 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10607956 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10607958 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1426269_PM_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1452007_PM_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10607952 |
- |
- |
Mouse Genome 430 2.0 Array |
1452007_at |
- |
- |
Mouse Genome 430 2.0 Array |
1426269_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1452007_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1426269_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000018.mm.1 |
NM_011515.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000018.mm.1 |
TCX_GL456233_random00000005.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000020.mm.1 |
NM_011515.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000020.mm.1 |
TCX_GL456233_random00000005.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000024.mm.1 |
NM_011515.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000024.mm.1 |
TCX_GL456233_random00000005.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000022.mm.1 |
NM_011515.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000022.mm.1 |
TCX_GL456233_random00000005.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000023.mm.1 |
NM_011515.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000023.mm.1 |
TCX_GL456233_random00000005.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000019.mm.1 |
NM_011515.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000019.mm.1 |
TCX_GL456233_random00000005.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000021.mm.1 |
NM_011515.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000021.mm.1 |
TCX_GL456233_random00000005.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000025.mm.1 |
NM_011515.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSRX_GL456233_random00000025.mm.1 |
TCX_GL456233_random00000005.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1452007_at |
- |
- |
Mouse Expression Set 430 |
1452007_PM_at |
- |
- |
Mouse Expression Set 430 |
1426269_PM_at |
- |
- |
Mouse Expression Set 430 |
1426269_at |
- |
- |
Murine Genome U74v2 Set |
162113_PM_r_at |
- |
- |
Murine Genome U74v2 Set |
102885_PM_at |
- |
- |
Murine Genome U74v2 Set |
162113_r_at |
- |
- |
Murine Genome U74v2 Set |
102885_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1426269_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1452007_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1452007_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1426269_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1426269_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1452007_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1452007_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1426269_PM_at |
- |
- |