Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnmbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY383729.1 | - | 6 - 7 | 2744 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |