Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181564.2 | NP_853543.1 | 13 - 14 | 2220 | 78 | |
GenBank mRNA | AB240192.1 | - | 12 - 13 | 2403 | 78 | |
AY191584.1 | - | 13 - 14 | 2220 | 78 | ||
AY227349.1 | - | 13 - 14 | 2086 | 78 | ||
BC109009.2 | - | 13 - 14 | 2220 | 78 | ||
BC150475.2 | - | 13 - 14 | 2220 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |