Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183021.3 | NP_898842.2 | 28 - 29 | 4213 | 64 | |
GenBank mRNA | AK173258.1 | - | 13 - 14 | 1723 | 64 | |
BC052885.1 | - | 3 - 4 | 353 | 64 | ||
EF222207.1 | - | 28 - 29 | 4121 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |