Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168600.1 | NP_001162071.1 | 13 - 14 | 1443 | 63 | |
NM_198110.2 | NP_932778.1 | 13 - 14 | 1476 | 63 | ||
GenBank mRNA | BC020354.1 | - | 13 - 14 | 1412 | 63 | |
BC057033.1 | - | 13 - 14 | 1402 | 6834283 | 63 | |
BC079653.1 | - | 13 - 14 | 1398 | 6843880 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |