Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168600.1 | NP_001162071.1 | 9 - 10 | 897 | 56 | |
NM_198110.2 | NP_932778.1 | 9 - 10 | 930 | 56 | ||
GenBank mRNA | BC020354.1 | - | 9 - 10 | 866 | 56 | |
BC057033.1 | - | 9 - 10 | 856 | 6834283 | 56 | |
BC079653.1 | - | 9 - 10 | 852 | 6843880 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |