Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168600.1 | NP_001162071.1 | 10 - 11 | 986 | 79 | |
NM_198110.2 | NP_932778.1 | 10 - 11 | 1019 | 79 | ||
GenBank mRNA | BC020354.1 | - | 10 - 11 | 955 | 79 | |
BC057033.1 | - | 10 - 11 | 945 | 6834283 | 79 | |
BC079653.1 | - | 10 - 11 | 941 | 6843880 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |