Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25996047 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25996049 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25996048 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P495471 |
BC031460.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P495471 |
BC031460.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
264170 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
859202 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
217351 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
666702 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
301536 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
6170 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
611489 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
35546 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
965041 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
187056 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
526447 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
165133 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
329281 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
680263 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
885367 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
449565 |
NM_001081052.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
355898 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
783045 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
946701 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
403048 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
741878 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
156434 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
663177 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911960 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911107 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
867241 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
616416 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
725040 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
305335 |
BC002118.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
74556 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
117087 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
2565 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
115762 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
85710 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P168985 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P111475 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P495471 |
BC031460.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P495471 |
BC031460.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P495471 |
BC031460.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P495471 |
BC031460.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P194765 |
BC031460.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P194765 |
NM_026904.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P194765 |
AK005303.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P194765 |
AK041373.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25996053 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25996062 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25996059 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25996068 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25996047 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25996061 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25996064 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25996063 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25996049 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25996051 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25996056 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25996058 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25996067 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P168985 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P111475 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06568058 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01776343 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01776330 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06568002 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06568045 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06568021 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P194765 |
BC031460.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P194765 |
NM_026904.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P194765 |
AK005303.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P194765 |
AK041373.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P495471 |
BC031460.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P495471 |
BC031460.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P109920 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P111475 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P01776330 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P168985 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P06568045 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07101035 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07101030 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07101034 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25996048 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25996049 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25996047 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996284 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996309 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996212 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996292 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996247 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996311 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996092 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996123 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996215 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996074 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996338 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996336 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996272 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996152 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996226 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996100 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996335 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996131 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996179 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996224 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996154 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996181 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996204 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996165 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996120 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996235 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996133 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996267 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996283 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996098 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996053 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996240 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996257 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996113 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996102 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996329 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996059 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996075 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996104 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996217 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996328 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996049 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996174 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996219 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996237 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996242 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996170 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996313 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996301 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996047 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996221 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996126 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996296 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996230 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996156 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996262 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996064 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996323 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996245 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996095 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996071 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996115 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996085 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996161 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996234 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996157 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996180 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996304 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996210 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996275 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996130 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996207 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996117 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996214 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996077 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996195 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996251 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996299 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996084 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996091 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996051 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996200 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996183 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996246 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996249 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996176 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996193 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996209 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996190 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996205 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996206 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996280 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996122 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996243 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996088 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996270 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996134 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996061 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996056 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996254 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996148 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996228 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996138 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996151 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996238 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996264 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996197 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996231 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996136 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996057 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996327 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25996158 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07280492 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996231 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996242 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996258 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996053 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996176 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996226 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996154 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06881128 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07073705 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996061 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996246 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996122 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996202 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996120 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996165 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996077 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996170 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996079 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996063 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996166 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996230 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07066839 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06909434 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06881133 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996051 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996238 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996251 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996174 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07185598 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996123 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996059 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996064 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996075 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996212 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996209 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996237 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996084 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996080 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996168 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996083 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996206 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996161 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996068 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07112247 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996224 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996200 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996056 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996172 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996047 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07181755 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06705510 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996126 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996234 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996235 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996243 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07201779 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996082 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07112250 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996117 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06881134 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996128 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06881136 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996219 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996245 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996221 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996215 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07185600 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996247 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996071 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996115 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996257 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996062 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996127 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996249 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996074 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996254 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996259 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996240 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996152 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996217 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996049 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25996057 |
- |
- |