Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgsm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197943.2 | NP_922934.2 | 19 - 20 | 2756 | 79 | |
GenBank mRNA | AK122273.1 | - | 19 - 20 | 2678 | 79 | |
AK197195.1 | - | 1 - 2 | 56 | 79 | ||
AK199077.1 | - | 1 - 2 | 56 | 79 | ||
AK199338.1 | - | 1 - 2 | 56 | 79 | ||
AK199845.1 | - | 1 - 2 | 56 | 79 | ||
AK199979.1 | - | 1 - 2 | 56 | 79 | ||
AK203050.1 | - | 1 - 2 | 56 | 79 | ||
AK212740.1 | - | 1 - 2 | 56 | 79 | ||
AK213455.1 | - | 1 - 2 | 56 | 79 | ||
BC058414.1 | - | 19 - 20 | 2747 | 6822416 | 79 | |
BC060163.1 | - | 19 - 20 | 2747 | 6822416 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |