Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010149.3 | NP_034279.3 | 6 - 7 | 972 | 158 | |
GenBank mRNA | AK010968.1 | - | 5 - 6 | 680 | 158 | |
AK139031.1 | - | 6 - 7 | 871 | 158 | ||
BC003953.1 | - | 6 - 7 | 825 | 158 | ||
BC046282.1 | - | 6 - 7 | 902 | 6438591 | 158 | |
J04843.1 | - | 6 - 7 | 854 | 158 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |