Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC011217.1 | - | 3 - 4 | 968 | 4195356 | 208 |
BC016407.1 | - | 3 - 4 | 979 | 4210846 | 208 | |
BC019802.1 | - | 3 - 4 | 968 | 5050045 | 208 | |
D00725.1 | - | 969 | 208 | |||
X55147.1 | - | 3 - 4 | 974 | 208 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |