Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rprd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291134.1 | NP_001278063.1 | 4 - 5 | 920 | 70 | |
NM_001291135.1 | NP_001278064.1 | 4 - 5 | 920 | 70 | ||
NM_001291136.1 | NP_001278065.1 | 3 - 4 | 787 | 70 | ||
NM_027434.3 | NP_081710.1 | 4 - 5 | 917 | 70 | ||
GenBank mRNA | AK011979.1 | - | 4 - 5 | 918 | 70 | |
AK045404.1 | - | 3 - 4 | 593 | 70 | ||
AK139995.1 | - | 619 | 70 | |||
AK147718.1 | - | 4 - 5 | 890 | 70 | ||
AK147930.1 | - | 4 - 5 | 879 | 70 | ||
AK152125.1 | - | 4 - 5 | 718 | 70 | ||
AK152220.1 | - | 4 - 5 | 718 | 70 | ||
BC028819.1 | - | 4 - 5 | 702 | 5100766 | 70 | |
BC042447.1 | - | 4 - 5 | 629 | 5357316 | 70 | |
BC054350.1 | - | 4 - 5 | 620 | 3488763 | 70 | |
DQ372939.1 | - | 4 - 5 | 921 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |