Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10519742 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519741 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519650 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519740 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519739 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519743 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10519651 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1421668_PM_x_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10519738 |
- |
- |
Mouse Genome 430 2.0 Array |
1421668_PM_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1421668_x_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
71026_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
71026_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0500000785.mm.1 |
TC0500000091.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500000785.mm.1 |
NM_027650.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000785.mm.1 |
NM_027650.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000782.mm.1 |
TC0500000091.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500000782.mm.1 |
NM_027650.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000782.mm.1 |
NM_027650.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000783.mm.1 |
TC0500000091.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500000783.mm.1 |
NM_027650.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000783.mm.1 |
NM_027650.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000784.mm.1 |
TC0500000091.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500000784.mm.1 |
NM_027650.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000784.mm.1 |
NM_027650.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000786.mm.1 |
TC0500000091.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500000786.mm.1 |
NM_027650.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000786.mm.1 |
NM_027650.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000787.mm.1 |
TC0500000091.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500000787.mm.1 |
NM_027650.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500000787.mm.1 |
NM_027650.3 |
Refseq |
Mouse Expression Set 430 |
1421668_PM_x_at |
- |
- |
Mouse Expression Set 430 |
1421668_x_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1421668_x_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1421668_PM_x_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1421668_x_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1421668_PM_x_at |
- |
- |