Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031170.2 | NP_112447.2 | 4 - 5 | 811 | 213 | |
GenBank mRNA | AK077597.1 | - | 4 - 5 | 784 | 213 | |
AK145546.1 | - | 4 - 5 | 784 | 213 | ||
AK145679.1 | - | 4 - 5 | 784 | 213 | ||
AK145777.1 | - | 4 - 5 | 811 | 213 | ||
AK146948.1 | - | 4 - 5 | 781 | 213 | ||
AK147044.1 | - | 4 - 5 | 781 | 213 | ||
AK166737.1 | - | 4 - 5 | 784 | 213 | ||
AK166768.1 | - | 4 - 5 | 781 | 213 | ||
AK166854.1 | - | 4 - 5 | 784 | 213 | ||
AK166960.1 | - | 4 - 5 | 784 | 213 | ||
AK166993.1 | - | 4 - 5 | 806 | 213 | ||
AK167120.1 | - | 4 - 5 | 781 | 213 | ||
AK167269.1 | - | 4 - 5 | 784 | 213 | ||
AK167471.1 | - | 4 - 5 | 781 | 213 | ||
AK168522.1 | - | 4 - 5 | 781 | 213 | ||
AK168540.1 | - | 4 - 5 | 781 | 213 | ||
AK168561.1 | - | 4 - 5 | 781 | 213 | ||
AK168726.1 | - | 4 - 5 | 781 | 213 | ||
AK168910.1 | - | 4 - 5 | 781 | 213 | ||
AK169136.1 | - | 4 - 5 | 784 | 213 | ||
AK169691.1 | - | 4 - 5 | 782 | 213 | ||
BC094009.1 | - | 4 - 5 | 775 | 4987971 | 213 | |
M21836.1 | - | 4 - 5 | 772 | 213 | ||
M22831.1 | - | 4 - 5 | 743 | 213 | ||
X12789.1 | - | 4 - 5 | 1084 | 213 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |