Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080798.1 | NP_001074267.1 | 15 - 16 | 3124 | 70 | |
NM_133919.3 | NP_598680.3 | 15 - 16 | 3135 | 70 | ||
GenBank mRNA | AF013131.1 | - | 14 - 15 | 2950 | 70 | |
AF074266.1 | - | 15 - 16 | 2965 | 70 | ||
AY749168.1 | - | 15 - 16 | 2968 | 70 | ||
BC131665.1 | - | 15 - 16 | 2968 | 40110795 | 70 | |
CN537099.1 | - | 2 - 3 | 133 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |