Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500012731.mm.1 |
NM_153178.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012731.mm.1 |
NM_153178.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012731.mm.1 |
TC1500001689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012738.mm.1 |
NM_153178.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012738.mm.1 |
NM_153178.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012738.mm.1 |
TC1500001689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012733.mm.1 |
NM_153178.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012733.mm.1 |
NM_153178.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012733.mm.1 |
TC1500001689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012737.mm.1 |
NM_153178.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012737.mm.1 |
NM_153178.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012737.mm.1 |
TC1500001689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012734.mm.1 |
NM_153178.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012734.mm.1 |
NM_153178.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012734.mm.1 |
TC1500001689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012735.mm.1 |
NM_153178.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012735.mm.1 |
NM_153178.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012735.mm.1 |
TC1500001689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012730.mm.1 |
NM_153178.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012730.mm.1 |
NM_153178.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012730.mm.1 |
TC1500001689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012739.mm.1 |
NM_153178.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012739.mm.1 |
NM_153178.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012739.mm.1 |
TC1500001689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012740.mm.1 |
NM_153178.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012740.mm.1 |
NM_153178.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012740.mm.1 |
TC1500001689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012741.mm.1 |
NM_153178.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012741.mm.1 |
NM_153178.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012741.mm.1 |
TC1500001689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012732.mm.1 |
NM_153178.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012732.mm.1 |
NM_153178.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012732.mm.1 |
TC1500001689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012728.mm.1 |
NM_153178.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012728.mm.1 |
NM_153178.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012728.mm.1 |
TC1500001689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012729.mm.1 |
NM_153178.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012729.mm.1 |
NM_153178.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012729.mm.1 |
TC1500001689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500012736.mm.1 |
NM_153178.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012736.mm.1 |
NM_153178.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500012736.mm.1 |
TC1500001689.mm.1 |
MTA TC |