Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183390.3 | NP_899246.2 | 1 - 2 | 332 | 111 | |
GenBank mRNA | AY350736.1 | - | 1 - 2 | 260 | 111 | |
BC100568.1 | - | 1 - 2 | 319 | 30073940 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |