Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmr1nb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166619.1 | NP_001160091.1 | 3 - 4 | 564 | 108 | |
NM_001166620.1 | NP_001160092.1 | 2 - 3 | 486 | 108 | ||
NM_174993.2 | NP_778158.2 | 4 - 5 | 702 | 108 | ||
GenBank mRNA | AK076190.1 | - | 3 - 4 | 522 | 108 | |
AY214130.1 | - | 4 - 5 | 607 | 108 | ||
BC099411.1 | - | 3 - 4 | 564 | 30788714 | 108 | |
BC138087.1 | - | 3 - 4 | 545 | 8861106 | 108 | |
BC145139.1 | - | 4 - 5 | 683 | 9053689 | 108 | |
BC145140.1 | - | 4 - 5 | 683 | 9053690 | 108 | |
BC171922.1 | - | 2 - 3 | 467 | 9053688 | 108 | |
BC171923.1 | - | 3 - 4 | 545 | 9053691 | 108 | |
BC171947.1 | - | 4 - 5 | 683 | 9054233 | 108 | |
BC171948.1 | - | 4 - 5 | 683 | 9054234 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |