Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175638.3 | NP_783569.1 | 11 - 12 | 2249 | 83 | |
GenBank mRNA | AY184228.1 | - | 11 - 12 | 2218 | 83 | |
AY187027.1 | - | 11 - 12 | 2174 | 83 | ||
AY260059.1 | - | 11 - 12 | 2212 | 83 | ||
BC026591.1 | - | 2 - 3 | 188 | 83 | ||
BC026679.1 | - | 4 - 5 | 448 | 83 | ||
BC037114.1 | - | 11 - 12 | 2209 | 83 | ||
BC043677.1 | - | 11 - 12 | 1622 | 83 | ||
BC096453.1 | - | 11 - 12 | 2205 | 4234311 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |