Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175638.3 | NP_783569.1 | 12 - 13 | 2389 | 120 | |
GenBank mRNA | AY184228.1 | - | 12 - 13 | 2358 | 120 | |
AY187027.1 | - | 12 - 13 | 2314 | 120 | ||
AY260059.1 | - | 12 - 13 | 2352 | 117 | ||
BC026591.1 | - | 3 - 4 | 328 | 120 | ||
BC026679.1 | - | 5 - 6 | 588 | 120 | ||
BC043677.1 | - | 12 - 13 | 1762 | 120 | ||
BC096453.1 | - | 12 - 13 | 2345 | 4234311 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |