Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10548516 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10548528 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10548518 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10548517 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10548521 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10548526 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10548531 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10548519 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10548520 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10548529 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10548527 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10548530 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10548525 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10548513 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0600025596.mm.1 |
TC0600003612.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025596.mm.1 |
NM_008459.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025596.mm.1 |
NM_008459.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025598.mm.1 |
TC0600003612.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025598.mm.1 |
NM_008459.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025598.mm.1 |
NM_008459.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025599.mm.1 |
TC0600003612.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025599.mm.1 |
NM_008459.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025599.mm.1 |
NM_008459.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025597.mm.1 |
TC0600003612.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025597.mm.1 |
NM_008459.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025597.mm.1 |
NM_008459.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025600.mm.1 |
TC0600003612.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025600.mm.1 |
NM_008459.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025600.mm.1 |
NM_008459.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025601.mm.1 |
TC0600003612.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025601.mm.1 |
NM_008459.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025601.mm.1 |
NM_008459.2 |
Refseq |