Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp33b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK009327.2 | - | 1 | 452 | 140 | |
AK049668.1 | - | 796 | 140 | |||
BC046593.1 | - | 1 | 617 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon |