Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24182862 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24182787 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24182785 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24182860 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24182789 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24182788 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24182859 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24182786 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24182861 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2032708 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2025153 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2082986 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2134251 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2002689 |
Z11997.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2002689 |
BC111468.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2002689 |
BP760493.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2002689 |
BC008565.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2002689 |
AK157846.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2013501 |
Z11997.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2013501 |
BC111468.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2013501 |
BC094030.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2013501 |
BC100312.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2013501 |
BC110667.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2018283 |
BC008565.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2018283 |
AK157846.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2018283 |
AK146420.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2018283 |
AK084841.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2018283 |
AK034535.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
345204 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
333889 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
457564 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
81332 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2134251 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2002689 |
Z11997.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2002689 |
BC111468.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2002689 |
BP760493.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2002689 |
BC008565.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2002689 |
AK157846.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2032708 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2013501 |
Z11997.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2013501 |
BC111468.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2013501 |
BC094030.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2013501 |
BC100312.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2013501 |
BC110667.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2025153 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2018283 |
BC008565.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2018283 |
AK157846.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2018283 |
AK146420.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2018283 |
AK084841.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2018283 |
AK034535.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2082986 |
- |
- |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P323044 |
- |
- |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P269554 |
Z11997.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P269554 |
BC111468.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P269554 |
BP760493.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P269554 |
BC008565.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P269554 |
AK157846.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P324336 |
BC064790.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182984 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182992 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182979 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182969 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182982 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182963 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182996 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182981 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182972 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182986 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182990 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182980 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182970 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182975 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182959 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182997 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182964 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182988 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182968 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24183000 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182966 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182978 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24182994 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01249337 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05787500 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P117821 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P323044 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P269554 |
Z11997.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P269554 |
BC111468.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P269554 |
BP760493.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P269554 |
BC008565.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P269554 |
AK157846.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P324336 |
BC064790.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P104715 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P114727 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P101783 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P139509 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P105048 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P113837 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P106442 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P105810 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P130997 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P133579 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P117815 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04631019 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04889933 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04683924 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24182786 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24182785 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24182862 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24182859 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24182789 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24182861 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24182788 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24182860 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24182787 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182924 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182989 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182890 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182969 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182903 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182972 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182978 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182998 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182904 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182979 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182954 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24183004 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24183015 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182917 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182892 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182929 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182930 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24183014 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182984 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182982 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182896 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182997 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182996 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182994 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24183019 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182907 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182963 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182946 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24183010 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182950 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182905 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182949 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24183008 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182915 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182898 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24183020 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182888 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182922 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182981 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182920 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182976 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182952 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182980 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182966 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24183021 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24183005 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24183011 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182928 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182908 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182968 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182932 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24182914 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182963 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182892 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182989 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182988 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04228097 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182932 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04683924 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182886 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04683918 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04788911 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182929 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182998 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04344159 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182994 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182894 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04747849 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04889933 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04117561 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182956 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04788917 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182911 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182996 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04117555 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182959 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24183000 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24183001 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182893 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182946 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24183008 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182980 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182992 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182957 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182915 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182917 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182982 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182949 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04629791 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04575834 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04103649 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04788915 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182950 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04328881 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04786062 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04683920 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182952 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182888 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04228095 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182930 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P24182945 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04273549 |
- |
- |