Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10607439 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10607438 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10607435 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10607434 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10607456 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10607445 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10607455 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10607443 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10607454 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10607437 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10607433 |
- |
- |
Mouse Genome 430 2.0 Array |
1450542_s_at |
- |
- |
Mouse Genome 430 2.0 Array |
1450542_PM_s_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0X00020205.mm.1 |
NM_020017.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020205.mm.1 |
NM_020017.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020205.mm.1 |
TC0X00003228.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020207.mm.1 |
NM_020017.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020207.mm.1 |
NM_020017.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020207.mm.1 |
TC0X00003228.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020206.mm.1 |
NM_020017.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020206.mm.1 |
NM_020017.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020206.mm.1 |
TC0X00003228.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1450542_PM_s_at |
- |
- |
Mouse Expression Set 430 |
1450542_s_at |
- |
- |
Murine Genome U74v2 Set |
103291_PM_at |
- |
- |
Murine Genome U74v2 Set |
103291_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1450542_s_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1450542_PM_s_at |
- |
- |