Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027427.2 | NP_081703.1 | 7 - 8 | 692 | 121 | |
GenBank mRNA | AK051568.1 | - | 7 - 8 | 693 | 121 | |
AK146331.1 | - | 7 - 8 | 655 | 121 | ||
AK166550.1 | - | 7 - 8 | 685 | 121 | ||
BC137591.1 | - | 7 - 8 | 666 | 8860610 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |