Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009483.1 | NP_033509.1 | 6 - 7 | 902 | 108 | |
GenBank mRNA | AJ002730.1 | - | 5 - 6 | 361 | 108 | |
AK080370.1 | - | 6 - 7 | 921 | 108 | ||
AK164941.1 | - | 6 - 7 | 934 | 108 | ||
BC053433.1 | - | 6 - 7 | 902 | 6516368 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |