Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem63b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198167.3 | NP_937810.2 | 20 - 21 | 2074 | 82 | |
GenBank mRNA | AK090200.1 | - | 20 - 21 | 1995 | 82 | |
AK159673.1 | - | 20 - 21 | 2079 | 82 | ||
BC026370.1 | - | 3 - 4 | 333 | 82 | ||
BC039277.1 | - | 9 - 10 | 1056 | 82 | ||
BC057136.1 | - | 9 - 10 | 1037 | 6529222 | 82 | |
BC059283.1 | - | 13 - 14 | 1422 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |