Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040403.1 | NP_001035493.1 | 2 - 3 | 254 | 62 | |
GenBank mRNA | AK004589.1 | - | 2 - 3 | 192 | 62 | |
AK030199.1 | - | 2 - 3 | 217 | 62 | ||
AK170557.1 | - | 2 - 3 | 255 | 62 | ||
AK213895.1 | - | 2 - 3 | 109 | 62 | ||
BY195482.1 | - | 2 - 3 | 255 | 62 | ||
BY204171.1 | - | 2 - 3 | 255 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |