Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011262.5 | NP_035392.1 | 6 - 7 | 730 | 62 | |
GenBank mRNA | AK004812.1 | - | 6 - 7 | 667 | 62 | |
AK138047.1 | - | 6 - 7 | 664 | 62 | ||
AK144106.1 | - | 6 - 7 | 669 | 62 | ||
AK144848.1 | - | 6 - 7 | 665 | 62 | ||
AK144954.1 | - | 6 - 7 | 676 | 62 | ||
AK145851.1 | - | 6 - 7 | 726 | 62 | ||
AK156252.1 | - | 5 - 6 | 500 | 62 | ||
BC007188.1 | - | 5 - 6 | 512 | 62 | ||
BC012709.1 | - | 6 - 7 | 660 | 4208205 | 62 | |
U10435.1 | - | 6 - 7 | 617 | 62 | ||
U43921.1 | - | 6 - 7 | 682 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |