Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard3nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK002760.1 | - | 4 - 5 | 523 | 70 | |
AK018331.1 | - | 5 - 6 | 524 | 70 | ||
AK152116.1 | - | 5 - 6 | 586 | 70 | ||
AK152378.1 | - | 5 - 6 | 537 | 70 | ||
BC003334.1 | - | 5 - 6 | 540 | 3256725 | 70 | |
BG084215.2 | - | 5 - 6 | 542 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |