Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600000595.mm.1 |
TC0600000078.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000595.mm.1 |
NM_001168382.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000595.mm.1 |
NM_001168382.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000595.mm.1 |
NM_029404.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000595.mm.1 |
NM_029404.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000600.mm.1 |
TC0600000078.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000600.mm.1 |
NM_029404.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000600.mm.1 |
NM_029404.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000601.mm.1 |
TC0600000078.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000601.mm.1 |
NM_029404.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000601.mm.1 |
NM_029404.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000602.mm.1 |
TC0600000078.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000602.mm.1 |
NM_029404.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000602.mm.1 |
NM_029404.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000597.mm.1 |
TC0600000078.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000596.mm.1 |
TC0600000078.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000596.mm.1 |
NM_001168382.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000596.mm.1 |
NM_001168382.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000596.mm.1 |
NM_029404.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000596.mm.1 |
NM_029404.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000598.mm.1 |
TC0600000078.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000598.mm.1 |
NM_029404.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000598.mm.1 |
NM_029404.3 |
Refseq |