Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600000585.mm.1 |
TC0600000078.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000585.mm.1 |
NM_001168382.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000585.mm.1 |
NM_001168382.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000585.mm.1 |
NM_029404.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000585.mm.1 |
NM_029404.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000583.mm.1 |
TC0600000078.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000583.mm.1 |
NM_001168382.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000583.mm.1 |
NM_001168382.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000583.mm.1 |
NM_029404.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000583.mm.1 |
NM_029404.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000584.mm.1 |
TC0600000078.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000584.mm.1 |
NM_001168382.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000584.mm.1 |
NM_001168382.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000584.mm.1 |
NM_029404.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000584.mm.1 |
NM_029404.3 |
Refseq |