Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286062.1 | NP_001272991.1 | 2 - 3 | 970 | 65 | |
NM_009640.4 | NP_033770.2 | 2 - 3 | 970 | 65 | ||
GenBank mRNA | AK012027.1 | - | 2 - 3 | 906 | 65 | |
AK088439.1 | - | 2 - 3 | 971 | 65 | ||
AK172868.1 | - | 2 - 3 | 790 | 65 | ||
BC067410.1 | - | 2 - 3 | 788 | 30436666 | 65 | |
U83509.1 | - | 2 - 3 | 760 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |