Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cav1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007616.4 | NP_031642.1 | 1 - 2 | 146 | 68 | |
GenBank mRNA | AK003489.1 | - | 1 - 2 | 94 | 68 | |
AK028738.1 | - | 1 - 2 | 80 | 68 | ||
AK035592.1 | - | 1 - 2 | 84 | 68 | ||
AY439333.1 | - | 1 - 2 | 65 | 68 | ||
BC038280.1 | - | 1 - 2 | 54 | 4484857 | 68 | |
BC052859.1 | - | 1 - 2 | 57 | 30062454 | 68 | |
BY235137.1 | - | 1 - 2 | 121 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |