Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023729.3 | NP_076218.3 | 3 - 4 | 346 | 78 | |
GenBank mRNA | AF459789.1 | - | 3 - 4 | 340 | 78 | |
AK016595.1 | - | 3 - 4 | 341 | 78 | ||
BC058859.1 | - | 3 - 4 | 355 | 30011651 | 78 | |
BG088136.2 | - | 3 - 4 | 346 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |