Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290651.1 | NP_001277580.1 | 3 - 4 | 613 | 92 | |
NM_001290652.1 | NP_001277581.1 | 3 - 4 | 613 | 92 | ||
GenBank mRNA | AK016533.1 | - | 6 - 7 | 485 | 92 | |
AK031490.1 | - | 3 - 4 | 592 | 92 | ||
AK040521.1 | - | 3 - 4 | 615 | 92 | ||
BC056382.1 | - | 3 - 4 | 303 | 6853936 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |