Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290651.1 | NP_001277580.1 | 7 - 8 | 1267 | 94 | |
NM_001290652.1 | NP_001277581.1 | 7 - 8 | 1267 | 94 | ||
GenBank mRNA | AF285577.1 | - | 9 - 10 | 1093 | 94 | |
AK016533.1 | - | 10 - 11 | 1139 | 94 | ||
AK031490.1 | - | 7 - 8 | 1246 | 94 | ||
AK040521.1 | - | 7 - 8 | 1269 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |