Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mboat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153546.4 | NP_705774.1 | 6 - 7 | 698 | 100 | |
GenBank mRNA | AB297382.1 | - | 6 - 7 | 521 | 100 | |
AB305045.1 | - | 6 - 7 | 521 | 100 | ||
AK033388.1 | - | 6 - 7 | 640 | 100 | ||
AK137755.1 | - | 6 - 7 | 630 | 100 | ||
AK146984.1 | - | 6 - 7 | 605 | 100 | ||
BC023845.1 | - | 6 - 7 | 702 | 5346000 | 100 | |
BC024653.1 | - | 6 - 7 | 650 | 3660479 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |