Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081116.1 | NP_001074585.1 | 6 - 7 | 4234 | 102 | |
GenBank mRNA | AK122250.1 | - | 6 - 7 | 889 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |