Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcdc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195617.1 | NP_001182546.1 | 3 - 4 | 693 | 93 | |
NM_177577.3 | NP_808245.2 | 3 - 4 | 693 | 93 | ||
GenBank mRNA | AK220370.1 | - | 3 - 4 | 466 | 93 | |
BC045136.1 | - | 3 - 4 | 693 | 6306251 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |